missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, ≥99.9%, Solstice
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.7822 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | 6°C |
| Packaging | Glass Bottle |
| UN Number | UN1648 |
| Water | 0.01% max. |
| Beilstein | 741857 |
| Solubility Information | Miscible in Water at 20°C |
| Boiling Point | 82°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile |