missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, ≥99.9%, Solstice

Catalog No. 6000272
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000272 4 x 4 L Glass Bottle
6000274 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000272 Supplier Solstice Supplier No. 0154

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Quantity 4 x 4 L
Flash Point 6°C
Packaging Glass Bottle
UN Number UN1648
Water 0.01% max.
Beilstein 741857
Solubility Information Miscible in Water at 20°C
Boiling Point 82°C
Formula Weight 41
Percent Purity >99.9%
Grade HPLC Gradient
Chemical Name or Material Acetonitrile, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, ≥99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile