missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, B&J Brand™, for applications requiring low aldehydes & ketones, carbonyl-free, meets UV Specifications, ≥99.9%, Honeywell Burdick & Jackson
$1376.93 - $2675.60
Chemical Identifiers
| CAS | 75-05-8 |
|---|---|
| Molecular Formula | C2H3N |
| Molecular Weight (g/mol) | 41.053 |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| IUPAC Name | acetonitrile |
| SMILES | CC#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6000229
|
Honeywell-Burdick and Jackson
0181L |
6 x 1 L | Glass Bottle |
Case of 6 Each for $1,376.93
|
|
||||
|
6000228
|
Honeywell-Burdick and Jackson
0184 |
4 x 4 L | Glass Bottle |
Case of 4 Each for $2,675.60
|
|
||||
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
| C2H3N | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
Specifications
| 75-05-8 | |
| 100.0000% | |
| 82°C | |
| 6°C | |
| C2H3N | |
| 0.01% max. | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41 | |
| Acetonitrile, B&J Brand™, for applications requiring low aldehydes & ketones, carbonyl-free, meets UV Specifications, ≥99.9% |
| 0.05 AU max. at 200 nm (UV), 0.04 AU max. at 210 nm (UV) | |
| 0.7822 g/cm3 | |
| 6 x 1 L | |
| Glass Bottle | |
| UN1648 | |
| 741857 | |
| Miscible in Water at 20°C | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| >99.9% |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile