missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for applications requiring extremely low water content, meets UV Specifications, ≥99.9%, Solstice
Specifications
Specifications
| Absorbance | 0.05 AU max. at 200 nm (UV), 0.04 AU max. at 210 nm (UV), 0.01 AU max. at 225 nm (UV), 0.005 AU max. at 250 nm (UV), 0.005 AU max. at 350 nm (UV) |
| CAS Max % | 100.0000% |
| Density | 0.7822 g/cm3 |
| Quantity | 4 x 4 L |
| Flash Point | 6°C |
| Packaging | Glass Bottle |
| Water | 0.003% max. |
| UN Number | UN1648 |
| Beilstein | 741857 |
| Boiling Point | 82°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile |