missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, anhydrous, amine free, 99.9%

CAS: 75-05-8 | C2H3N | 41.053 g/mol

Supplier:  Thermo Scientific Chemicals 043440K7

Catalog No. AA43440K7


Only null left
Explore available promotions

Description

Description

In peptide and DNA synthesis, HPLC.Acetonitrile is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. It is used as a mobile phase in HPLC and in LC-MS. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetonitrile
Amine Free
Colorless
80°C to 82°C
5°C (41°F)
Pungent
1.344
C2H3N
73mm Hg at 20°C
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
-48°C to -45°C
75-05-8
0.786 g/cm3 at 20°C
4 L
525°C (977°F)
Liquid
99.9%
MFCD00001878
741857
Miscible with water and methanol.
CC#N
41.053
CHEBI:38472

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Keep away from sources of ignition - No Smoking. Wear suitable protective clothing and gloves.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c

H225-H302+H312+H332-H319

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only