missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA43166K2
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43166K2 1 L
AA43166AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43166K2 Supplier Thermo Scientific Chemicals Supplier No. 043166K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-05-8 | C2H3N | 41.053 g/mol

Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point °C to 48 to°C to 45°C
Density 0.786
Quantity 1 L
Flash Point 5°C (41°F)
Physical Form Liquid
Refractive Index 1.344
Assay Percent Range >99.8%
Beilstein 741857
Boiling Point 80°C to 82°C
Solubility Information Miscible with water,alcohols,ethers,acetone,chloroform,carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions).
Formula Weight 41.05
Chemical Name or Material Acetonitrile, anhydrous, Water <100ppm
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 200-835-2
RTECSNumber AL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only