missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Honeywell

Catalog No. p-7112042
Change view
Click to view available options
Quantity:
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000285 4 x 4 L Glass Bottle
6000286 4 x 4 L Poly Bottle
6000284 204 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000285 Supplier Honeywell Chemicals Supplier No. LP0144
Only null left

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Boiling Point 82°C
Quantity 4 x 4 L
Flash Point 6°C
Packaging Glass Bottle
UN Number UN1648
Beilstein 741857
Water 0.3% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 41
Percent Purity >99.5%
Grade ACS Reagent
Assay by GC Analysis
Chemical Name or Material Acetonitrile, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile