missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 99%

Catalog No. p-7024703
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Catalog No. Quantity
AAA19862AP 500 mL
AAA198620F 2500 mL
2 options

Catalog No. AAA19862AP

Supplier: Thermo Scientific Chemicals A19862AP

Only null left

CAS: 75-05-8 | C2H3N | 41.053 g/mol

Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

Solubility
Miscible with water, alcohols, ethers, acetone, chloroform, carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions).

Notes
Incompatible with alkali metals, acids, bases, reducing agents and oxidizing agents. Highly flammable. Store it away from heat, sparks, and flame exposure.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Density 0.786
Boiling Point 80°C to 82°C
Flash Point 5°C (41°F)
Odor Pungent
Refractive Index 1.344
Linear Formula CH3CN
Quantity 500 mL
UN Number UN1648
Beilstein 741857
Merck Index 14,70
Solubility Information Miscible with water,alcohols,ethers,acetone,chloroform,carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions).
Formula Weight 41.05
Percent Purity 99%
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
EINECSNumber 200-835-2
RTECSNumber AL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only