missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, For pesticide residue analysis, Honeywell Riedel-de Haën™
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
| Melting Point | -48°C (literature) |
| CAS Max % | 100.0000% |
| Density | 0.786 g/mL at 25 °C(lit.) |
| Quantity | 4 x 2.5 L |
| Flash Point | 6°C |
| Packaging | Glass Bottle |
| Linear Formula | CH3CN |
| UN Number | UN1648 |
| Beilstein | 741857 |
| Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
| Show More |
Safety and Handling
| ShelfLife | 720 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile |
| EINECSNumber | 200-835-2 |
| Recommended Storage | Room Temp |