missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ≥99.5% (GC), Honeywell™

≥99.5% (GC)

Supplier:  Honeywell Chemicals 600044X5L

 View more versions of this product

Catalog No. 60047150


Only null left
Explore available promotions

Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Specifications

Acetonitrile
75-05-8
0.786g/mL at 25°C
4 x 5 L
973°C
1.344
CH3CN
72.8mmHg (20°C)
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05g/mol
-48°C
Colorless
81°C to 82°C
0.16
Poly Bottle
C2H3N
MFCD00001878
UN1648
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.5% (GC)

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338

DOTInformation : 3 / PGII

RTECSNumber : 200-835-2

SDS
Documents

Documents

Promotions

Promotions