missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, ≥99.5% (GC), Honeywell™
≥99.5% (GC)
Supplier: Honeywell Chemicals 6000425L
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
| Acetonitrile | |
| 75-05-8 | |
| Colorless liquid | |
| 81.6°C | |
| 16% | |
| Poly Bottle | |
| CH3CN | |
| 72.8mmHg (20°C) | |
| 741857 | |
| 1000g/L at 25°C | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| ≥99.5% (GC) |
| -45.7°C | |
| Colorless | |
| 0.786g/mL at 25°C | |
| 2.5 L | |
| 973°C | |
| C2H3N | |
| MFCD00001878 | |
| UN1648 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05g/mol |
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
Safety and Handling
P210-P280-P305 + P351 + P338
DOTInformation : 3 / PGII
RTECSNumber : 200-835-2