missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Acetonitrile

Catalog No. 7334664L
Encompass
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7334664L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7334664L Supplier Sigma Aldrich Supplier No. 7334664L

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acetonitrile

Specifications

CAS 75-05-8
Melting Point -45°C
Density 0.786 g/mL (at 25°C)
Boiling Point 81°C to 82°C
Molecular Formula C2H3N
Refractive Index n20/D 1.344 (literature)
Linear Formula CH3CN
Quantity 4 L
MDL Number MFCD00001878
Synonym ACN; Cyanomethane; Ethyl nitrile; Methyl cyanide
Molecular Weight (g/mol) 41.05
Percent Purity 99.999% trace metals basis
Show More Show Less