missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, GR, ACS, MilliporeSigma™

Catalog No. 50947651
Change view
Click to view available options
Quantity:
20 L
Packaging:
Steel Barrel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50947651 20 L Steel Barrel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947651 Supplier Technidata Supplier No. AX01493

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Color Colorless
Appearance Clear
Density 0.786g/cc at 20°C
Quantity 20 L
Flash Point 2°C
Ignition Point 524°C
Odor Ether-Like
Packaging Steel Barrel
Titratable Acid >8μeq/g
Vapor Pressure 97 hPa (20°C)
Viscosity 0.316 mPa.s at 25°C
Water >0.3%
UN Number UN 1648
Boiling Point 81.6°C (1013 hPa)
Formula Weight 41.05
Percent Purity <99.5%
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category Danger
Hazard Statement H225: Highly flammable liquid and vapor. H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled. H319: Causes serious eye irritation.
Precautionary Statement P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P240 Ground/bond container and receiving equipment. P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P403 + P233 Store in a well-ventilated place. Keep container tightly closed
EINECSNumber 200-835-2
RTECSNumber AL7700000
Recommended Storage Store at room temperature.