missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 18% v/v, Aqueous, Buffered with 1% Sodium Acetate, Reagents

Catalog No. 01335970
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335970 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335970 Supplier Labchem Inc Supplier No. CA1006004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetonitrile, 18% v/v, Aqueous, Buffered with 1% Sodium Acetate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetonitrile, 18% v/v, Aqueous, Buffered with 1% Sodium Acetate
CAS 7732-18-5, 75-05-8, 75-05-8
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9651
Molecular Formula C2H3N/H2O
Synonym Methyl cyanide
Boiling Point 76°C
Percent Purity 18%
Grade Reagent
Show More Show Less