missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, Honeywell Burdick & Jackson
Specifications
Specifications
| Boiling Point | 82°C |
| CAS Min % | >99.0000%, 0.0350% |
| CAS Max % | >90%, <0.1000% |
| UN Number | UN1648 |
| Density | 0.777 g/cm3 |
| Grade | Solvent Blend |
| Packaging | Hdpe Nowpak™ |
| Quantity | 20 L |
| Flash Point | 6°C |
| Chemical Name or Material | Acetonitrile with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution |