missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% trifluoroacetic acid, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS, LabReady™ Solvent Blend
$5987.75 - $5987.75
Chemical Identifiers
| CAS | 75-05-8 |
|---|---|
| Molecular Formula | C2H3N |
| MDL Number | MFCD00001878 |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| IUPAC Name | acetonitrile |
| SMILES | CC#N |
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Chemical Identifiers
| 75-05-8 | |
| MFCD00001878 | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| CHEBI:38472 | |
| CC#N |
Specifications
| 75-05-8 | |
| 6°C (42.8°F) | |
| C2H3N | |
| UN1648 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| >99.5% (GC) |
| 4 x 4 L | |
| Glass Bottle | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile | |
| CC#N | |
| 41.05 | |
| CHEBI:38472 | |
| Acetonitrile with 0.1% trifluoroacetic acid |
Safety and Handling
P210-P305 + P351 + P338
Danger
EINECSNumber : 200-835-2