missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade

Catalog No. LS120212
Change view
Click to view available options
Quantity:
500mL
1 L
2.5 L
4L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LS120212 2.5 L Glass Bottle
LS120500 500mL Glass Bottle
LS1201 1 L Wide Neck Amber Bottle
LS1204 4L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LS120212 Supplier Thermo Fisher Scientific Supplier No. LS120 212

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, formisoton, formylic acid, collo-bueglatt, formira, aminic acid, bilorin, acide formique, myrmicyl, hydrogen carboxylic acid, methanoic acid, 2.5L | CAS: 75-05-8

0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 1L

  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of the resulting separation

Chemical Identifiers

CAS 75-05-8

Specifications

Grade LC-MS
Item Name 0.1% Formic Acid in Acetonitrile (v/v)
Quantity 2.5 L
Form Liquid
Reagent Type Solvent Blends
Packaging Glass Bottle
LCMS Gradient Suitability Pass Test