missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Solstice
Specifications
Specifications
| Boiling Point | 82°C |
| CAS Min % | >99.0000% |
| CAS Max % | >90%, 0.1000% |
| Density | 0.7767 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | 8°C |
| Packaging | Glass Bottle, Gl45 |
| UN Number | UN1648 |
| Grade | HPLC |
| Chemical Name or Material | Acetonitrile with 0.1% Formic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution |