missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.1 ammonium acetate Tested for UHPLC-MS, MilliporeSigma™ Supelco™

Catalog No. 111018115
Change view
Click to view available options
Quantity:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018115 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018115 Supplier Merck Emd Millipore Supplier No. 142742L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00001878

Specifications

CAS 148642-19-7
Quantity 2 L
Flash Point 5°C
ShelfLife Limited shelf life, expiry date on the label