missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.05% Trifluoroacetic Acid (TFA), Optima LC/MS, Fisher Chemical

Catalog No. LS1174
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LS1174 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LS1174 Supplier Thermo Fisher Scientific Supplier No. LS1174
Get 1 Extra Chemical When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal high purity organic mobile phase for LC/MS and UHPLC-UV applications, specifically protein and peptide separations

  • Suitable for LC-MS and UHPLC-UV
  • Lower TFA content enhances separations by minimizing ion suppression
  • Pre-blended mobile phase simplifies preparation leaving more time to run and analyze samples
  • TFA is a common mobile phase additive and ion pair reagent in chromatography
  • Complementary to the 0.05% TFA aqueous blend -- LS1154

Chemical Identifiers

CAS 76-05-1

Specifications

Grade OPTIMA LC-MS
Quantity 4 L
Form Liquid