missing translation for 'onlineSavingsMsg'
Learn More

Acetoin (May exist as crystalline dimer) 98.0+%, TCI America™

Catalog No. p-7130423
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H022525G 25 g
H0225500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H022525G Supplier TCI America Supplier No. H022525G
Only null left

Chemical Identifiers

CAS 513-86-0
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00004521,MFCD00038696
InChI Key ROWKJAVDOGWPAT-UHFFFAOYNA-N
Synonym acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
PubChem CID 179
ChEBI CHEBI:15688
IUPAC Name 3-hydroxybutan-2-one
SMILES CC(O)C(C)=O

Specifications

Color White-Yellow
Boiling Point 144°C
Quantity 25 g
UN Number 2621
Formula Weight 88.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acetoin (May exist as crystalline dimer)
EINECSNumber (2)-0597
RTECSNumber EL8790000
TSCA Yes
Recommended Storage Refrigerator