missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetohydroxamic Acid 98.0+%, TCI America™
$92.41 - $248.34
Chemical Identifiers
| CAS | 546-88-3 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.067 |
| MDL Number | MFCD00009994 |
| InChI Key | RRUDCFGSUDOHDG-UHFFFAOYSA-N |
| Synonym | acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure |
| PubChem CID | 1990 |
| ChEBI | CHEBI:49029 |
| IUPAC Name | N-hydroxyacetamide |
| SMILES | CC(=O)NO |
Chemical Identifiers
| 546-88-3 | |
| 75.067 | |
| RRUDCFGSUDOHDG-UHFFFAOYSA-N | |
| 1990 | |
| N-hydroxyacetamide |
| C2H5NO2 | |
| MFCD00009994 | |
| acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure | |
| CHEBI:49029 | |
| CC(=O)NO |
Specifications
| 546-88-3 | |
| White-Yellow | |
| MFCD00009994 | |
| acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure | |
| CC(=O)NO | |
| 75.067 | |
| CHEBI:49029 | |
| ≥98.0% (T) | |
| Acetohydroxamic Acid |
| 92°C | |
| C2H5NO2 | |
| 10 g | |
| RRUDCFGSUDOHDG-UHFFFAOYSA-N | |
| N-hydroxyacetamide | |
| 1990 | |
| 75.07 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : AK8157000
TSCA : No