missing translation for 'onlineSavingsMsg'
Learn More

Acetohydroxamic Acid 98.0+%, TCI America™

Catalog No. A005125G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A005125G 25 g
A005110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A005125G Supplier TCI America Supplier No. A005125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 546-88-3
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.067
MDL Number MFCD00009994
InChI Key RRUDCFGSUDOHDG-UHFFFAOYSA-N
Synonym acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure
PubChem CID 1990
ChEBI CHEBI:49029
IUPAC Name N-hydroxyacetamide
SMILES CC(=O)NO

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Formula Weight 75.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetohydroxamic Acid
RTECSNumber AK8157000
TSCA No