missing translation for 'onlineSavingsMsg'
Learn More

Acetohydroxamic acid, 98+%

Catalog No. AC170130250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170130250 25 g Glass bottle
AC170130050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170130250 Supplier Thermo Scientific Chemicals Supplier No. 170130250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 546-88-3 | C2H5NO2 | 75.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 546-88-3
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.06
MDL Number MFCD00009994
InChI Key RRUDCFGSUDOHDG-UHFFFAOYSA-N
Synonym acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure
PubChem CID 1990
ChEBI CHEBI:49029
IUPAC Name N-hydroxyacetamide
SMILES CC(=O)NO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 86.0°C to 90.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3C(=NOH)OH
Quantity 25 g
Beilstein 02, 187
Fieser 13,2
Merck Index 15, 62
Solubility Information Solubility in water: soluble
Formula Weight 75.06
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material Acetohydroxamic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May damage the unborn child.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-913-8

RUO – Research Use Only