missing translation for 'onlineSavingsMsg'
Learn More

Acetobromo-alpha-D-glucuronic acid methyl ester, 98%

CAS: 21085-72-3 | C13H17BrO9 | 397.17 g/mol

$363.51 - $363.51

Chemical Identifiers

CAS 21085-72-3
Molecular Formula C13H17BrO9
Molecular Weight (g/mol) 397.17
InChI Key GWTNLHGTLIBHHZ-SVNGYHJRSA-N
Synonym 2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, a-acetobromoglucuronate, bromomethyltriacetyl glucuronate, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, methyl acetobromo-alpha-d-glucuronate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, methyl, methyl
PubChem CID 88785
IUPAC Name methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
SMILES CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC436290010
SDS View Documents
Thermo Scientific Chemicals
436290010
1 g Glass bottle
Each for $363.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21085-72-3
397.17
2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, a-acetobromoglucuronate, bromomethyltriacetyl glucuronate, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, methyl acetobromo-alpha-d-glucuronate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, methyl, methyl
methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
C13H17BrO9
GWTNLHGTLIBHHZ-SVNGYHJRSA-N
88785
CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C

Specifications

21085-72-3
100.0
White to Gray
97.5% min. (ELSD) (HPLC)
C13H17BrO9
+185° to +195° (20°C, 589nm) (c=1, ethyl acetate)
GWTNLHGTLIBHHZ-SVNGYHJRSA-N
methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
2% (CaCO3)
397.17
Crystalline Powder or Solid
97.5
80°C to 110°C
Authentic
Glass bottle
1 g
2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, a-acetobromoglucuronate, bromomethyltriacetyl glucuronate, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, methyl acetobromo-alpha-d-glucuronate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, methyl, methyl
CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C
397.17
88785
98%
Acetobromo-α-D-glucuronic acid methyl ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 244-203-4

SDS
Documents

Documents

RUO – Research Use Only