missing translation for 'onlineSavingsMsg'
Learn More

Acetobromo-alpha-D-glucuronic acid methyl ester, 98%

Catalog No. AC436290010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436290010 1 g Glass bottle
AC436290050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436290010 Supplier Thermo Scientific Chemicals Supplier No. 436290010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21085-72-3 | C13H17BrO9 | 397.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21085-72-3
Molecular Formula C13H17BrO9
Molecular Weight (g/mol) 397.17
InChI Key GWTNLHGTLIBHHZ-SVNGYHJRSA-N
Synonym 2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, acetobromo-alpha-d-glucuronic acid methyl ester, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, methyl acetobromo-alpha-d-glucuronate, 2,3,4-tri-o-acetyl-alpha-d-glucopyranosyl bromide uronic acid methyl ester, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, acetobromo-alpha-d-glucuronic acid, methyl ester, methyl, a-acetobromoglucuronate
PubChem CID 88785
IUPAC Name methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
SMILES CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 80°C to 110°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation +185° to +195° (20°C, 589nm) (c=1, ethyl acetate)
Stabilizer 2% (CaCO3)
Formula Weight 397.17
Percent Purity 98%
Physical Form Crystalline Powder or Solid
Chemical Name or Material Acetobromo-α-D-glucuronic acid methyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 244-203-4

RUO – Research Use Only