missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide 99.0+%, TCI America™

Catalog No. A0046500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0046500G 500 g
A004625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0046500G Supplier TCI America Supplier No. A0046500G

Chemical Identifiers

CAS 102-01-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00008780
InChI Key DYRDKSSFIWVSNM-UHFFFAOYSA-N
Synonym acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
PubChem CID 7592
IUPAC Name 3-oxo-N-phenylbutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 500 g
Formula Weight 177.20
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Acetoacetanilide
EINECSNumber (3)-0110
RTECSNumber AK4200000
TSCA Yes