missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide, 98+%

Catalog No. AAA1753430
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1753430 250 g
AAA175340B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1753430 Supplier Thermo Scientific Chemicals Supplier No. A1753430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-01-2 | C10H11NO2 | 177.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-01-2
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD00008780
InChI Key DYRDKSSFIWVSNM-UHFFFAOYSA-N
Synonym acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
PubChem CID 7592
IUPAC Name 3-oxo-N-phenylbutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 83°C to 86°C
Density 1.26
Boiling Point 129°C (24 mmHg)
Flash Point 150°C (302°F)
Odor Odorless
Quantity 250 g
Beilstein 473419
Merck Index 14,58
Formula Weight 177.2
Percent Purity ≥98%
Chemical Name or Material Acetoacetanilide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280f
Wear protective clothing.
EINECSNumber 202-996-4
RTECSNumber AK4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only