missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, for HPLC, Thermo Scientific Chemicals
GREENER_CHOICE

Catalog No. p-7214368
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446202500 250 g
AC446200010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446202500 Supplier Thermo Scientific Chemicals Supplier No. 446202500
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
IUPAC Name sodium acetate trihydrate
SMILES O.O.O.[Na+].CC([O-])=O

Specifications

Melting Point 58.0°C
Linear Formula CH3COONa·3 H2O
Merck Index 15, 8709
Solubility Information Solubility aq. soln.: 762 g/L (20°C). Other solubilities: 1 g/19 mL alcohol
Formula Weight 136.08
Grade HPLC
Quantity 250 g
Green Features ACT Ecolabel (Gold)
Chemical Name or Material Acetic acid, sodium salt trihydrate

RUO – Research Use Only