missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, sodium salt trihydrate, 99+%, extra pure
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
$122.58 - $122.58
Chemical Identifiers
| CAS | 127-09-3 |
|---|---|
| Molecular Formula | C2H3NaO2 |
| Molecular Weight (g/mol) | 82.03 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
| PubChem CID | 517045 |
| ChEBI | CHEBI:32954 |
| SMILES | [Na+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC123240010
|
Thermo Scientific Chemicals
123240010 |
1 kg | Plastic Bottle |
Each for $122.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| [Na+].CC([O-])=O |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 |
Specifications
| 58.0°C | |
| 99.0 | |
| White | |
| C2H3NaO2 | |
| MFCD00012459 | |
| 01,1024; 05,591 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium;acetate | |
| 517045 | |
| 136.08 | |
| Extra Pure | |
| Authentic | |
| Adhering Crystals |
| 127-09-3,6131-90-4 | |
| 100.0 | |
| 99+% | |
| CH3COONa·3H2O | |
| 02, 96 | |
| 15, 8709 | |
| Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| ≥99% | |
| 1 kg | |
| Plastic Bottle | |
| Acetic acid, sodium salt trihydrate |
RUO – Research Use Only