missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, 99+%, for biochemistry

Catalog No. p-4380035
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446792500 250 g Plastic bottle
AC446790010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446792500 Supplier Thermo Scientific Chemicals Supplier No. 446792500
Only null left

CAS: 6131-90-4 | C2H3NaO2·3H2O | 136.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H3NaO2·3H2O
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Absorbance 0.05 max. (3M) at 260nm, 0.05 max. (3M) at 280nm
Melting Point 58.0°C
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3COONa·3H2O
Quantity 250 g
Merck Index 15,8709
Solubility Information Solubility in water: 762g/L (20°C). Other solubilities: 1g/19mL alcohol
Formula Weight 136.08
Percent Purity 99+%
Grade Biochemistry
Insoluble Matter 0.005% max. (in water)
Chemical Name or Material Acetic acid, sodium salt trihydrate
Show More Show Less

RUO – Research Use Only