missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, sodium salt trihydrate, 99+%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Absorbance | 0.05 max. (3M) at 260nm, 0.05 max. (3M) at 280nm |
| Melting Point | 58.0°C |
| Assay Percent Range | 99+% |
| Packaging | Plastic bottle |
| Linear Formula | CH3COONa·3H2O |
| Quantity | 1 kg |
| Merck Index | 15,8709 |
| Solubility Information | Solubility in water: 762g/L (20°C). Other solubilities: 1g/19mL alcohol |
| Formula Weight | 136.08 |
| Percent Purity | 99+% |
| Show More |
RUO – Research Use Only