missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, 99+%, ACS reagent

Catalog No. AC424260010
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424260010 1 kg Plastic bottle
AC424260250 25 g Plastic bottle
AC424260025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424260010 Supplier Thermo Scientific Chemicals Supplier No. 424260010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-09-3 , 6131-90-4
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

pH 7.5 to 9.2 (5% soln. at 25°C)
CAS Min % 99.0
Color White
CAS Max % 100.0
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline
Assay Percent Range 99+%
Linear Formula CH3COONa·3H2O
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
IUPAC Name sodium acetate
Formula Weight 136.08
Percent Purity 99.0 to 101.0%
Grade ACS Reagent
Chemical Name or Material Acetic acid, sodium salt trihydrate
Show More Show Less
Hazard Category HYGROSCOPIC
RTECSNumber AJ4580000

RUO – Research Use Only