missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, 99.5%, for analysis

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Supplier:  Thermo Scientific Chemicals 217110010

 View more versions of this product

Catalog No. AC217110010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Specifications

Acetic acid, sodium salt trihydrate
p.a.
127-09-3,6131-90-4
100.0
99.5%
CH3COONa·3H2O
02, 96
15, 8709
Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol
[Na+].CC([O-])=O
82.03
CHEBI:32954
99.5%
1 kg
Plastic bottle
7.5 to 9.0 (5 % at 20°C)
58.0°C
99.4
White
C2H3NaO2
MFCD00012459
01,1024; 05,591
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
517045
136.08
Analytical
Authentic
Crystalline

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
sodium acetate
C2H3NaO2
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
CHEBI:32954
[Na+].CC([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

RTECSNumber : AJ4580000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only