missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Absorbance | (c= 0.1 M, water), (c= 0.1 M, water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm |
| Melting Point | 324.0°C |
| Assay Percent Range | 99% |
| Linear Formula | CH3CO2Na |
| Solubility Information | Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol |
| Formula Weight | 82.03 |
| Percent Purity | 99% |
| Grade | Biochemical |
| Quantity | 250 g |
| Flash Point | >250°C |
| Show More |
RUO – Research Use Only