missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid/Isooctane, 60:40, Ricca Chemical

Catalog No. R01610004C1
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01610004C1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01610004C1 Supplier Ricca Chemical Company Supplier No. R01610004C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 64-19-7, 540-84-1
CAS Min % 38.0, 62.0
CAS Max % 42.0, 58.0
Quantity 4 L
Odor Vinegar/gasoline-like odor
Packaging Safety-coated Glass Bottle
Physical Form Liquid
Molecular Formula C2H4O2
MDL Number MFCD00036152
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) Mixture
PubChem CID 176
ChEBI CHEBI:15366
Grade Laboratory
Chemical Name or Material Acetic Acid/ Isooctane
Show More Show Less