missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid/Hexane Solution, Reagents

Catalog No. 01335905
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335905 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335905 Supplier Labchem Inc Supplier No. CA2043004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid/Hexane Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetic Acid/Hexane Solution
CAS 110-54-3, 64-19-7
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.6642
Synonym Ethanoic Acid/Hexane
Grade Reagent