missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 98%, pure

CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol

$1028.04 - $1028.04

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169880100
SDS View Documents
Thermo Scientific Chemicals
169880100
10 kg Plastic drum
Each for $1,028.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

631-61-8
77.08
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate

Specifications

631-61-8
White
Authentic
Plastic drum
CH3CO2NH4
02,96
15,492
Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
CC(=O)[O-].[NH4+]
77.08
CHEBI:62947
98%
Adhering Crystals
114.0°C
136°C
98%
C2H7NO2
10 kg
01,38; 07,11
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.08
Pure
Acetic acid, ammonium salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-162-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only