missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 98%, pure

Catalog No. AC169880010
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169880010 1 kg Plastic Bottle
AC169880025 2.5 kg Plastic Bottle
AC169880100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169880010 Supplier Thermo Scientific Chemicals Supplier No. 169880010

CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 114.0°C
Color White
Flash Point 136°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Linear Formula CH3CO2NH4
Quantity 1 kg
Beilstein 02,96
Fieser 01,38; 07,11
Merck Index 15,492
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity 98%
Grade Pure
Physical Form Adhering Crystals
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less
EINECSNumber 211-162-9

RUO – Research Use Only