missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetic acid, ammonium salt, 98%, for biochemistry

Catalog No. p-4440192
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447121000 100 g Plastic bottle
AC447125000 500 g Plastic bottle
AC447120025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC447121000 Supplier Thermo Scientific Chemicals Supplier No. 447121000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Absorbance (c=1M, water), (c=1M, water) at 0nm, 0.01 max. at 280nm, 0.01 max. at 350nm, 0.02 max. at 254nm
Melting Point 114.0°C
Assay Percent Range 98%
Linear Formula CH3CO2NH4
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity 98%
Grade Biochemical
Quantity 100 g
Flash Point 136°C
Packaging Plastic bottle
Chemical Name or Material Acetic acid, ammonium salt
Show More Show Less

RUO – Research Use Only