missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid-Ammonium Acetate Buffer TS, (U.S.P. Test Solution), Spectrum™ Chemical
Description
Spectrum™ Chemical Acetic Acid-Ammonium Acetate Buffer TS, (U.S.P. Test Solution) is a solution, sometimes designated with TS or Test Solution, that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
Specifications
Specifications
| CAS | 64-19-7, 7732-18-5 |
| CAS Max % | 0.1%, 99.9% |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Chemical Solution |
| Grade | USP |