missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid-Ammonium Acetate Buffer TS, (U.S.P. Test Solution), Spectrum™ Chemical

Catalog No. 18600237
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18600237 1 L
18600238 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600237 Supplier Spectrum Chemical Mfg Cor Supplier No. A2841LT
Only null left

A-284, 64-19-7

Spectrum™ Chemical Acetic Acid-Ammonium Acetate Buffer TS, (U.S.P. Test Solution) is a solution, sometimes designated with TS or Test Solution, that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Specifications

CAS 64-19-7, 7732-18-5
CAS Max % 0.1%, 99.9%
Grade USP
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Chemical Solution