missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer pH 4.6 to 4.7, For Bromide, Certified, LabChem™

Catalog No. LC100102
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC100102 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC100102 Supplier LabChem Supplier No. LC100102
Only null left

Specifications

Chemical Name or Material Acetate Buffer pH 4.6 to 4.7
Name Note For Bromide
CAS 7732-18-5, 7647-14-5, 6131-90-4, 64-19-7, 1310-73-2
Color Colorless
CAS Max % 80.7, 9, 6.8, 3, 0.5
Decomposition Information Carbon monoxide, Carbon dioxide
pH 4.6 to 4.7
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula CH3COOHNa·3H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.08
Grade Certified
Show More Show Less