Learn More
Thermo Scientific Chemicals Acetate Buffer (1 M, pH 5.5)
Description
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.
Notes
Contains sodium acetate and glacial acetic acid, pH 5.5
Specifications
Specifications
| Concentration | 1M buffer soln. |
| Molecular Formula | C2H3NaO2 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| SMILES | [Na+].CC([O-])=O |
| IUPAC Name | sodium acetate |
| Molecular Weight (g/mol) | 82.03 |
| Form | Liquid |
| Quantity | 250mL |
| Storage Requirements | Ambient temperatures |
RUO – Research Use Only