missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 5.5)

Catalog No. AAJ61033AP
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61033AP 500mL
AAJ61033AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61033AP Supplier Thermo Scientific Chemicals Supplier No. J61033AP

missing translation for 'validMainframeMsg'

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetate, 1M buffer soln., pH, 5.5 it is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.

Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.

Notes
Contains sodium acetate and glacial acetic acid, pH 5.5

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1M buffer soln.
Form Liquid
Quantity 500mL
Storage Requirements Ambient temperatures

RUO – Research Use Only