missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 5.0, Low Endotoxin)

Catalog No. AAJ67248AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67248AE 100 mL
AAJ67248AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67248AE Supplier Thermo Scientific Chemicals Supplier No. J67248AE

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.034
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Color Colorless
Concentration 1M buffer soln.
pH 5.0
Physical Form Liquid
Quantity 100 mL
Recommended Storage Ambient Temperatures

RUO – Research Use Only