missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 4.0)

Catalog No. AAJ60104AK
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60104AK 250mL
AAJ60104AP 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60104AK Supplier Thermo Scientific Chemicals Supplier No. J60104AK

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. Other reported uses. Used in the purification and precipitation of nucleic acids; Used in protein crystallization. Staining of gels in protein gel electrophoresis and HPLC; Mordant dyeing. DNA microarray studies of E. coli response; Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents and as a substrate of acetokinase.

Solubility
Fully miscible with water. Soluble in ethanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1M buffer soln.
Molecular Formula C2H3NaO2
pH Range 4
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
Form Liquid
Quantity 250mL
Storage Requirements Ambient temperatures
Show More Show Less

RUO – Research Use Only