missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetate Buffer (1 M, pH 3.5)

Catalog No. AAJ60340AP
Change view
Click to view available options
Quantity:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60340AP 500mL
AAJ60340AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60340AP Supplier Thermo Scientific Chemicals Supplier No. J60340AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Acetate, 1M buffer soln., pH, 3.5 is used in dexoyribionucleic acid (DNA) and ribonucleic acid (RNA) precipitation. It is also used in the preparation of gel stains for protein gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetate, 1M buffer soln., pH, 3.5 is used in dexoyribionucleic acid (DNA) and ribonucleic acid (RNA) precipitation. It is also used in the preparation of gel stains for protein gel electrophoresis.

Solubility
Soluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 127-09-3

Specifications

Concentration 1M buffer soln.
Form Liquid
Quantity 500mL
Storage Requirements Ambient temperatures

RUO – Research Use Only