missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acetate Buffer (1 M, pH 3.0)
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Supplier: Thermo Scientific Chemicals J63876AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsFor precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Acetate buffers for amine coupling of proteins to Biacore sensor chip surfaces. Studies on microbial pili suggest that sodium acetate suppressed K99 production in E. coli strains cultured on a minimal medium. Experiments have shown that when added to food waste composting systems, NaOAc may be effective at counteracting the adverse effects of organic acids produced in the composting process.
Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from water, moisture.
Specifications
| 127-09-3 | |
| C2H3NaO2 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium acetate | |
| Liquid | |
| Ambient temperatures |
| 1M buffer soln. | |
| MFCD00012459 | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| 250mL |
Chemical Identifiers
| 127-09-3 |
RUO – Research Use Only