missing translation for 'onlineSavingsMsg'
Learn More

Acetamidomethanol 96.0+%, TCI America™

Catalog No. A136925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A136925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A136925G Supplier TCI America Supplier No. A136925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-51-4
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00014417
InChI Key HWJHZLJIIWOTGZ-UHFFFAOYSA-N
Synonym n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol
PubChem CID 69365
IUPAC Name N-(hydroxymethyl)acetamide
SMILES CC(=O)NCO

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 25 g
Formula Weight 89.09
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acetamidomethanol
RTECSNumber AC3610000
TSCA No